Merck
  • USP15 Deubiquitinase Safeguards Hematopoiesis and Genome Integrity in Hematopoietic Stem Cells and Leukemia Cells.
USP15 Deubiquitinase Safeguards Hematopoiesis and Genome Integrity in Hematopoietic Stem Cells and Leukemia Cells.
Cell reports (2020-12-31)
Paul van den Berk, Cesare Lancini, Carlos Company, Michela Serresi, Maria Pilar Sanchez-Bailon, Danielle Hulsman, Colin Pritchard, Ji-Ying Song, Matthias Jürgen Schmitt, Ellen Tanger, Oliver Popp, Philipp Mertins, Ivo J Huijbers, Heinz Jacobs, Maarten van Lohuizen, Gaetano Gargiulo, Elisabetta Citterio
ABSTRACT
Altering ubiquitination by disruption of deubiquitinating enzymes (DUBs) affects hematopoietic stem cell (HSC) maintenance. However, comprehensive knowledge of DUB function during hematopoiesis in vivo is lacking. Here, we systematically inactivate DUBs in mouse hematopoietic progenitors using in vivo small hairpin RNA (shRNA) screens. We find that multiple DUBs may be individually required for hematopoiesis and identify ubiquitin-specific protease 15 (USP15) as essential for HSC maintenance in vitro and in transplantations and Usp15 knockout (KO) mice in vivo. USP15 is highly expressed in human hematopoietic tissues and leukemias. USP15 depletion in murine progenitors and leukemia cells impairs in vitro expansion and increases genotoxic stress. In leukemia cells, USP15 interacts with and stabilizes FUS (fused in sarcoma), a known DNA repair factor, directly linking USP15 to the DNA damage response (DDR). Our study underscores the importance of DUBs in preserving normal hematopoiesis and uncovers USP15 as a critical DUB in safeguarding genome integrity in HSCs and leukemia cells.
Sigma-Aldrich
MISSION® pLKO.1-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
Sigma-Aldrich
MISSION® esiRNA, targeting human USP15
Sigma-Aldrich
Resazurin sodium salt, certified by the BSC
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, from mouse