Merck
  • A combined spectroscopic and crystallographic approach to probing drug-human serum albumin interactions.
A combined spectroscopic and crystallographic approach to probing drug-human serum albumin interactions.
Bioorganic & medicinal chemistry (2010-09-28)
David Buttar, Nicola Colclough, Stefan Gerhardt, Philip A MacFaul, Scott D Phillips, Alleyn Plowright, Paul Whittamore, Kin Tam, Klaus Maskos, Stefan Steinbacher, Holger Steuber
ABSTRACT
The displacement of probes that bind selectively to subdomains IIA or IIIA on human serum albumin (HSA) by competing compounds has been followed using fluorescence spectroscopy, and has therefore been used to assign a primary binding site for these compounds in the presence and absence of fatty acids. The crystal structures have also been solved for three compounds: a matched pair of carboxylic acids whose binding strength to HSA unexpectedly decreased as the lipophilicity increased; and a highly bound sulphonamide that appeared not to displace the probes in the displacement assay. The crystallography results support the findings from the fluorescence displacement assay. The results indicate that drug binding to subdomain IB might also be important location for certain compounds.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Ibuprofen
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Phenylbutazone
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Glyburide, meets USP testing specifications
Supelco
Tolbutamide, analytical standard
Sigma-Aldrich
Furosemide
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)