Merck
  • X-aptamers targeting Thy-1 membrane glycoprotein in pancreatic ductal adenocarcinoma.
X-aptamers targeting Thy-1 membrane glycoprotein in pancreatic ductal adenocarcinoma.
Biochimie (2020-11-27)
Hongyu Wang, Xin Li, Lisa A Lai, Teresa A Brentnall, David W Dawson, Kimberly A Kelly, Ru Chen, Sheng Pan
ABSTRACT
Modified DNA aptamers incorporated with amino-acid like side chains or drug-like ligands can offer unique advantages and enhance specificity as affinity ligands. Thy-1 membrane glycoprotein (THY1 or CD90) was previously identified as a biomarker candidate of neovasculature in pancreatic ductal adenocarcinoma (PDAC). The current study developed and evaluated modified DNA X-aptamers targeting THY1 in PDAC. The expression and glycosylation of THY1 in PDAC tumor tissues were assessed using immunohistochemistry and quantitative proteomics. Bead-based X-aptamer library that contains 108 different sequences was used to screen for high affinity THY1 X-aptamers. The sequences of the X-aptamers were analyzed with the next-generation sequencing. The affinities of the selected X-aptamers to THY1 were quantitatively evaluated with flow cytometry. Three high affinity THY1 X-aptamers, including XA-B217, XA-B216 and XA-A9, were selected after library screening and affinity binding evaluation. These three X-aptamers demonstrated a high binding affinity and specificity to THY1 protein and the THY1 expressing cell lines, using THY1 antibody as a comparison. The development of these X-aptamers provides highly specific and non-immunogenic affinity ligands for THY1 binding in the context of biomarker development and clinical applications. They could be further exploited to assist molecular imaging of PDAC targeting THY1.
Sigma-Aldrich
Phorbol 12,13-dibutyrate, ≥98% (TLC), powder