Merck
  • The use of nanovibration to discover specific and potent bioactive metabolites that stimulate osteogenic differentiation in mesenchymal stem cells.
The use of nanovibration to discover specific and potent bioactive metabolites that stimulate osteogenic differentiation in mesenchymal stem cells.
Science advances (2021-02-28)
Tom Hodgkinson, P Monica Tsimbouri, Virginia Llopis-Hernandez, Paul Campsie, David Scurr, Peter G Childs, David Phillips, Sam Donnelly, Julia A Wells, Fergal J O'Brien, Manuel Salmeron-Sanchez, Karl Burgess, Morgan Alexander, Massimo Vassalli, Richard O C Oreffo, Stuart Reid, David J France, Matthew J Dalby
ABSTRACT
Bioactive metabolites have wide-ranging biological activities and are a potential source of future research and therapeutic tools. Here, we use nanovibrational stimulation to induce osteogenic differentiation of mesenchymal stem cells, in the absence of off-target, nonosteogenic differentiation. We show that this differentiation method, which does not rely on the addition of exogenous growth factors to culture media, provides an artifact-free approach to identifying bioactive metabolites that specifically and potently induce osteogenesis. We first identify a highly specific metabolite, cholesterol sulfate, an endogenous steroid. Next, a screen of other small molecules with a similar steroid scaffold identified fludrocortisone acetate with both specific and highly potent osteogenic-inducing activity. Further, we implicate cytoskeletal contractility as a measure of osteogenic potency and cell stiffness as a measure of specificity. These findings demonstrate that physical principles can be used to identify bioactive metabolites and then enable optimization of metabolite potency can be optimized by examining structure-function relationships.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Canrenone, ≥97% (HPLC)
Sigma-Aldrich
Anti-Osteonectin Antibody, serum, Chemicon®
Sigma-Aldrich
Mifepristone, ≥98%