Merck
  • Subchronic administration of auranofin reduced amyloid-β plaque pathology in a transgenic APPNL-G-F/NL-G-F mouse model.
Subchronic administration of auranofin reduced amyloid-β plaque pathology in a transgenic APPNL-G-F/NL-G-F mouse model.
Brain research (2020-07-25)
Jolanta Upīte, Inga Kadish, Thomas van Groen, Baiba Jansone
ABSTRACT
Alzheimer's disease (AD) is the most common cause of dementia. Neuropathological processes, including the accumulation of amyloid-β (Aβ) plaques and neurofibrillary tangles, and neuroinflammation, lead to cognitive impairment at middle and eventually later stages of AD progression. Over the last decade, focused efforts have explored repurposed drug approaches for AD pathophysiological mechanisms. Recently, auranofin, an anti-inflammatory drug, was shown to have therapeutic potential in a number of diseases in addition to rheumatoid arthritis. Surprisingly, no data regarding the effects of auranofin on cognitive deficits in AD mice or the influence of auranofin on Aβ pathology and neuroinflammatory processes are available. In the present study, we used 14-month-old transgenic male APPNL-G-F/NL-G-F mice to assess the effects of subchronic administration of auranofin at low doses (1 and 5 mg/kg, intraperitoneal) on spatial memory, Aβ pathology and the expression of cortical and hippocampal proteins (glial fibrillary acidic protein (GFAP), ionized calcium binding adaptor molecule-1 (Iba-1)) and proteins related to synaptic plasticity (glutamic acid decarboxylase 67 (GAD67), homer proteins homologue-1 (Homer-1)). The data demonstrated that auranofin significantly decreased Aβ deposition in the hippocampus and the number of Aβ plaques in the cingulate cortex, but it did not have memory-enhancing effects or induce changes in the expression of the studied proteins. Our current results highlight the importance of considering further pre-clinical research to investigate the possible beneficial effects of auranofin on the other pathological aspects of AD.
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Auranofin, ≥98% (HPLC)