Merck
  • Small extracellular vesicles containing miR-486-5p promote angiogenesis after myocardial infarction in mice and nonhuman primates.
Small extracellular vesicles containing miR-486-5p promote angiogenesis after myocardial infarction in mice and nonhuman primates.
Science translational medicine (2021-03-12)
Qingju Li, Yinchuan Xu, Kaiqi Lv, Yingchao Wang, Zhiwei Zhong, Changchen Xiao, Keyang Zhu, Cheng Ni, Kan Wang, Minjian Kong, Xuebiao Li, Youqi Fan, Fengjiang Zhang, Qi Chen, Yi Li, Qian Li, Chengjia Liu, Jinyun Zhu, Shuhan Zhong, Jingyi Wang, Yongjian Chen, Jing Zhao, Dan Zhu, Rongrong Wu, Jinghai Chen, Wei Zhu, Hong Yu, Reza Ardehali, Jianyi Jay Zhang, Jian'an Wang, Xinyang Hu
ABSTRACT
Stem cell-derived small extracellular vesicles (sEVs) promote angiogenesis after myocardial infarction (MI). However, the components of sEVs that contribute to these effects and the safety and efficiency of engineered sEV treatment for MI remain unresolved. Here, we observed improved cardiac function, enhanced vascular density, and smaller infarct size in mice treated with the sEVs from hypoxia-preconditioned (HP) mesenchymal stem cells (MSCs) (HP-sEVs) than in mice treated with normoxia-preconditioned (N) MSCs (N-sEVs). MicroRNA profiling revealed a higher abundance of miR-486-5p in HP-sEVs than in N-sEVs, and miR-486-5p inactivation abolished the benefit of HP-sEV treatment, whereas miR-486-5p up-regulation enhanced the benefit of N-sEV treatment. Matrix metalloproteinase 19 (MMP19) abundance was lower in HP-sEV-treated than N-sEV-treated mouse hearts but was enriched in cardiac fibroblasts (CFs), and Mmp19 was identified as one of the target genes of miR-486-5p. Conditioned medium from CFs that overexpressed miR-486-5p or silenced MMP19 increased the angiogenic activity of endothelial cells; however, medium from CFs that simultaneously overexpressed Mmp19 and miR-486-5p abolished this effect. Mmp19 silencing in CFs reduced the cleavage of extracellular vascular endothelial growth factor (VEGF). Furthermore, miR-486-5p-overexpressing N-sEV treatment promoted angiogenesis and cardiac recovery without increasing arrhythmia complications in a nonhuman primate (NHP) MI model. Collectively, this study highlights the key role of sEV miR-486-5p in promoting cardiac angiogenesis via fibroblastic MMP19-VEGFA cleavage signaling. Delivery of miR-486-5p-engineered sEVs safely enhanced angiogenesis and cardiac function in an NHP MI model and may promote cardiac repair.
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies