Merck
  • Design and synthesis of a biotinylated probe of COX-2 inhibitor nimesulide analog JCC76.
Design and synthesis of a biotinylated probe of COX-2 inhibitor nimesulide analog JCC76.
Bioorganic & medicinal chemistry letters (2011-08-03)
Bo Zhong, Rati Lama, Kerri M Smith, Yan Xu, Bin Su
ABSTRACT
JCC76 is a derivative of cyclooxygenase-2(COX-2) selective inhibitor nimesulide and exhibits potent anti-breast cancer activity. It selectively induces apoptosis of Her2 positive breast cancer cells. However, the specific molecular targets of JCC76 still remain unclear, which significantly withdraw the further drug development of JCC76. To identify the molecular targets of JCC76, a six carbon linker and biotin conjugated JCC76 probe was designed and synthesized. The anti-proliferation activity of the probe and its analogs was evaluated.
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%