Merck
  • Structural requirements of flavonoids for the adipogenesis of 3T3-L1 cells.
Structural requirements of flavonoids for the adipogenesis of 3T3-L1 cells.
Bioorganic & medicinal chemistry (2011-04-16)
Hisashi Matsuda, Yuichiro Kogami, Seikou Nakamura, Tomomi Sugiyama, Tsubasa Ueno, Masayuki Yoshikawa
ABSTRACT
To search for a new class of antidiabetic compounds, effects of 44 flavonoids on the adipogenesis of 3T3-L1 cells were examined. Among them, 3,4',7-trimethylkaempferol, tetramethylkaempferol, and pentamethylquercetin concentration-dependently enhanced the accumulation of triglyceride, a marker of adipogenesis. With regard to structural requirements of flavonoids for the activity, it was fond that: (1) most flavonoids having hydroxy groups lacked the effect; (2) flavonols with methoxy groups showed stronger effects particularly those with a methoxy group at the 3-position; and (3) a methoxy group of flavonols at the B ring was also important. 3,4',7-Trimethylkaempferol, tetramethylkaempferol, and pentamethylquercetin significantly increased the amount of adiponectin released into the medium and the uptake of 2-deoxyglucose into the cells. Furthermore, tetramethylkaempferol and pentamethylquercetin also increased mRNA levels of adiponectin, glucose transporter 4 (GLUT4), and fatty acid-binding protein (aP2). Both compounds also increased the mRNA levels of peroxisome proliferator-activated receptor (PPAR)γ2 and CCAAT/enhancer-binding protein (C/EBP)α, β, and/or δ, although, different from troglitazone, they did not activate PPARγ directly in a nuclear receptor cofactor assay.
Product Description
Supelco
(−)-Epigallocatechin, analytical standard
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Flavone, ≥99.0%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Isoquercitrin, primary reference standard
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Supelco
Quercetin 3-glucoside, analytical standard