Merck
Identification of novel ASK1 inhibitors using virtual screening.
Bioorganic & medicinal chemistry (2010-12-08)
Masako Okamoto, Nae Saito, Hirotatsu Kojima, Takayoshi Okabe, Kohsuke Takeda, Hidenori Ichijo, Toshio Furuya, Tetsuo Nagano
ABSTRACT
Apoptosis signal-regulating kinase 1 (ASK1, also called MAP3K5) is a mitogen-activated protein kinase kinase kinase (MAP3K) that plays important roles in stress-induced cell death and inflammation, and is expected as a new therapeutic target for cancer, cardiovascular diseases, and neurodegenerative diseases. We identified novel ASK1 inhibitors by virtual screening from the public chemical library collected by Chemical Biology Research Initiative (CBRI) at the University of Tokyo.
Sigma-Aldrich
6-(Dimethylamino)purine, ≥98%