Merck
  • Tubulin photoaffinity labeling study with a plinabulin chemical probe possessing a biotin tag at the oxazole.
Tubulin photoaffinity labeling study with a plinabulin chemical probe possessing a biotin tag at the oxazole.
Bioorganic & medicinal chemistry (2010-11-26)
Yuri Yamazaki, Yui Kido, Koushi Hidaka, Hiroyuki Yasui, Yoshiaki Kiso, Fumika Yakushiji, Yoshio Hayashi
ABSTRACT
A new bioactive photoaffinity probe KPU-252-B1 (4) possessing a biotin tag on the oxazole ring of a potent plinabulin derivative KPU-244 (2) was synthesized via the Cu(I)-catalyzed Huisgen's cycloaddition reaction to understand the precise binding mode of the diketopiperazine-based anti-microtubule agent plinabulin on tubulin. Probe 4 showed significant binding affinity toward tubulin and cytotoxicity against an HT-29 cells. A photoaffinity labeling study suggested that probe 4 specifically recognizes tubulin at a binding site that binds plinabulin or colchicine, most likely near or at the colchicine binding site, which is located at the interfacial region formed by α-and β-tubulin association. The results also demonstrated that probe 4 may serve as a useful plinabulin chemical probe to investigate the molecular mechanism by which anti-microtubule diketopiperazine derivatives operate.
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder