Merck
  • Isoflavone dimers and other bioactive constituents from the figs of Ficus mucuso.
Isoflavone dimers and other bioactive constituents from the figs of Ficus mucuso.
Journal of natural products (2011-05-31)
Jean J K Bankeu, Rustamova Khayala, Bruno N Lenta, Didérot T Noungoué, Silvère A Ngouela, Sufyan A Mustafa, Khalid Asaad, Mohammed I Choudhary, Sean T Prigge, Ralphreed Hasanov, Augustin E Nkengfack, Etienne Tsamo, Muhammad S Ali
ABSTRACT
Phytochemical investigation of the figs of Ficus mucuso led to the isolation of three new isoflavone dimer derivatives, mucusisoflavones A-C (1-3), together with 16 known compounds. Some of the isolates were tested in vitro for their inhibitory properties toward β-glucuronidase and Plasmodium falciparum enoyl-ACP reductase (PfENR) enzymes. Compound 1 (IC₅₀) 0.68 μM) showed inhibitory activity on β-glucuronidase enzyme, while 3 (IC₅₀) 7.69 μM) exhibited a weak inhibitory activity against P. falciparum enoyl-ACP reductase (PfENR).
Product Description
Supelco
Melting point standard 182-184°C, analytical standard
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
p-Anisic acid, ≥99%, FG
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
4-Methoxybenzoic acid, ReagentPlus®, 99%