Merck
  • Lanostane and hopane triterpenes from the entomopathogenic fungus Hypocrella sp. BCC 14524.
Lanostane and hopane triterpenes from the entomopathogenic fungus Hypocrella sp. BCC 14524.
Journal of natural products (2011-10-15)
Masahiko Isaka, Panida Chinthanom, Malipan Sappan, Rungtiwa Chanthaket, J Jennifer Luangsa-ard, Samran Prabpai, Palangpon Kongsaeree
ABSTRACT
Seven new lanostane-type triterpenes, hypocrellols A-G (1-7), and six new hopane-type triterpenes, 7β,15α-dihydroxy-22(29)-hopene (8), 3β,7β-dihydroxy-22(29)-hopene (9), 3β-acetoxy-15α-hydroxy-22(29)-hopene (10), 3β,7β,15α,22-tetrahydroxyhopane (11), 3β-acetoxy-7β,15α,22-trihydroxyhopane (12), and 7β,15α,22-trihydroxyhopane (13), were isolated from the scale insect pathogenic fungus Hypocrella sp. BCC 14524. The structures of the new compounds were elucidated by analyses of the NMR spectroscopic and mass spectrometry data. The structure of 1 was confirmed by X-ray crystallography.
Product Description
Supelco
17α(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Supelco
17β(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)