Merck
  • Radical scavenging and antioxidant activities of isocoumarins and a phthalide from the endophytic fungus Colletotrichum sp.
Radical scavenging and antioxidant activities of isocoumarins and a phthalide from the endophytic fungus Colletotrichum sp.
Journal of natural products (2010-12-23)
Kamolchanok Tianpanich, Surasak Prachya, Suthep Wiyakrutta, Chulabhorn Mahidol, Somsak Ruchirawat, Prasat Kittakoop
ABSTRACT
Five known isocoumarins, monocerin (1), derivative 2, and fusarentin derivatives 3-5, and a new phthalide (6) were isolated from the endophytic fungus Colletotrichum sp. 2 selectively exhibited cytotoxic activity toward the HepG2 cell line. Compounds 2 and 4 scavenged 2,2-diphenyl-1-picrylhydrazyl (DPPH) free radicals (IC(50) values of 23.4 and 16.4 μM, respectively) and inhibited superoxide anion radical formation (IC(50) values of 52.6 and 4.3 μM, respectively). The C-7 hydroxyl group in 2 and 4 might be important for radical scavenging activities. Isocoumarins 1-3 and phthalide 6 showed potent antioxidant activity.
Sigma-Aldrich
Phthalide, 98%
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration), meets EP testing specifications