Merck
  • Antimycobacterial activities of 5-alkyl (or halo)-3'-substituted pyrimidine nucleoside analogs.
Antimycobacterial activities of 5-alkyl (or halo)-3'-substituted pyrimidine nucleoside analogs.
Bioorganic & medicinal chemistry letters (2011-12-20)
Naveen C Srivastav, Neeraj Shakya, Sudha Bhavanam, Amogh Agrawal, Chris Tse, Nancy Desroches, Dennis Y Kunimoto, Rakesh Kumar
ABSTRACT
Several 5-alkyl (or halo)-3'-azido (amino or halo) analogs of pyrimidine nucleosides have been synthesized and evaluated against Mycobacterium bovis, Mycobacterium tuberculosis and Mycobacterium avium. Among these compounds, 3'-azido-5-ethyl-2',3'-dideoxyuridine (3) was found to have significant antimycobacterial activities against M. bovis (MIC(50)=1μg/mL), M. tuberculosis (MIC(50)=10μg/mL) and M. avium (MIC(50)=10μg/mL).
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
D-Cycloserine, synthetic
Sigma-Aldrich
D-Cycloserine