Merck
  • A Non-Coding Disease Modifier of Pancreatic Agenesis Identified by Genetic Correction in a Patient-Derived iPSC Line.
A Non-Coding Disease Modifier of Pancreatic Agenesis Identified by Genetic Correction in a Patient-Derived iPSC Line.
Cell stem cell (2020-05-23)
Siddharth Kishore, Elisa De Franco, Fabian L Cardenas-Diaz, Lisa R Letourneau-Freiberg, May Sanyoura, Catherine Osorio-Quintero, Deborah L French, Siri Atma W Greeley, Andrew T Hattersley, Paul Gadue
ABSTRACT
GATA6 is a critical regulator of pancreatic development, with heterozygous mutations in this transcription factor being the most common cause of pancreatic agenesis. To study the variability in disease phenotype among individuals harboring these mutations, a patient-induced pluripotent stem cell model was used. Interestingly, GATA6 protein expression remained depressed in pancreatic progenitor cells even after correction of the coding mutation. Screening the regulatory regions of the GATA6 gene in these patient cells and 32 additional agenesis patients revealed a higher minor allele frequency of a SNP 3' of the GATA6 coding sequence. Introduction of this minor allele SNP by genome editing confirmed its functionality in depressing GATA6 expression and the efficiency of pancreas differentiation. This work highlights a possible genetic modifier contributing to pancreatic agenesis and demonstrates the usefulness of using patient-induced pluripotent stem cells for targeted discovery and validation of non-coding gene variants affecting gene expression and disease penetrance.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
γ-Secretase Inhibitor XX, This γ-secretase inhibitor, CAS 209984-56-5, is a cell-permeable dibenzazepine compound that lowers both brain and plasma Aβ40 levels by ~72% in Tg2576 mutant APP transgenic mouse model.
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder