Merck
  • Synthesis and biological evaluation of benzo[d]imidazole derivatives as potential anti-cancer agents.
Synthesis and biological evaluation of benzo[d]imidazole derivatives as potential anti-cancer agents.
Bioorganic & medicinal chemistry letters (2012-01-10)
Hamad M Alkahtani, Abdullahi Y Abbas, Shudong Wang
ABSTRACT
We herein report the synthesis, biological activity and structure-activity relationship of derivatives of 5,6-dichloro-1-β-D-ribofuranosylbenzimidazole and benzo[d]imidazole. A lead compound 6o demonstrates potent anti-proliferative activity and the ability to induce cancer cell apoptosis.
Sigma-Aldrich
5,6-Dimethylbenzimidazole, ≥99%
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside