Merck
  • The site of breast cancer metastases dictates their clonal composition and reversible transcriptomic profile.
The site of breast cancer metastases dictates their clonal composition and reversible transcriptomic profile.
Science advances (2021-07-09)
Jean Berthelet, Verena C Wimmer, Holly J Whitfield, Antonin Serrano, Thomas Boudier, Stefano Mangiola, Michal Merdas, Farrah El-Saafin, David Baloyan, Jordan Wilcox, Steven Wilcox, Adam C Parslow, Anthony T Papenfuss, Belinda Yeo, Matthias Ernst, Bhupinder Pal, Robin L Anderson, Melissa J Davis, Kelly L Rogers, Frédéric Hollande, Delphine Merino
ABSTRACT
Intratumoral heterogeneity is a driver of breast cancer progression, but the nature of the clonal interactive network involved in this process remains unclear. Here, we optimized the use of optical barcoding to visualize and characterize 31 cancer subclones in vivo. By mapping the clonal composition of thousands of metastases in two clinically relevant sites, the lungs and liver, we found that metastases were highly polyclonal in lungs but not in the liver. Furthermore, the transcriptome of the subclones varied according to their metastatic niche. We also identified a reversible niche-driven signature that was conserved in lung and liver metastases collected during patient autopsies. Among this signature, we found that the tumor necrosis factor-α pathway was up-regulated in lung compared to liver metastases, and inhibition of this pathway affected metastasis diversity. These results highlight that the cellular and molecular heterogeneity observed in metastases is largely dictated by the tumor microenvironment.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Roche
Insulin, human, recombinant (yeast)
Sigma-Aldrich
FGF-2 / basic FGF Protein, Human recombinant, The FGF-2 / basic FGF Protein, Human recombinant is available in a 25 µg format.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid