Merck
  • Camphorsulfonic acid catalysed facile tandem double Friedlander annulation protocol for the synthesis of phenoxy linked bisquinoline derivatives and discovery of antitubercular agents.
Camphorsulfonic acid catalysed facile tandem double Friedlander annulation protocol for the synthesis of phenoxy linked bisquinoline derivatives and discovery of antitubercular agents.
Bioorganic & medicinal chemistry letters (2012-01-27)
Nidhin Paul, Muthuchamy Murugavel, Shanmugam Muthusubramanian, Dharmarajan Sriram
ABSTRACT
A series of phenoxy linked bisquinoline derivatives were synthesised from the Friedlander annulation of 2-(4-acetylphenoxy)-1-aryl-1-ethanones with 2-aminobenzophenone in good yields using (±)-camphor-10-sulfonic acid (CSA) as the catalyst. These compounds were screened for their in vitro activity against Mycobacterium tuberculosis H37Rv (MTB) and among the 23 compounds screened, 2-(3-bromophenyl)-6-chloro-3-[4-(6-chloro-4-phenyl-2-quinolyl)phenoxy]-4-phenylquinoline (3q) and 2-(4-bromophenyl)-6-chloro-3-[4-(6-chloro-4-phenyl-2-quinolyl)phenoxy]-4-phenylquinoline (3o) were found to be the most active compounds with MIC of 1.1 and 2.2 μM against MTB. The cytotoxic effects against mouse fibroblasts (NIH 3T3) in vitro were evaluated for 3o and 3q, which displayed no toxic effects against mouse fibroblast cell line NIH 3T3.
Sigma-Aldrich
(+)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Sigma-Aldrich
(1S)-(+)-10-Camphorsulfonic acid, 99%
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Pyrazinecarboxamide
Sigma-Aldrich
(−)-Camphor-10-sulfonic acid, purum, ≥98.0% (T)
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
(1R)-(−)-10-Camphorsulfonic acid, 98%
Sigma-Aldrich
Camphor-10-sulfonic acid (β), 98%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)