Merck
  • pH Responsiveness of Multilayered Films and Membranes Made of Polysaccharides.
pH Responsiveness of Multilayered Films and Membranes Made of Polysaccharides.
Langmuir : the ACS journal of surfaces and colloids (2015-10-01)
Joana M Silva, Sofia G Caridade, Rui R Costa, Natália M Alves, Thomas Groth, Catherine Picart, Rui L Reis, João F Mano
ABSTRACT
We investigated the pH-dependent properties of multilayered films made of chitosan (CHI) and alginate (ALG) and focused on their postassembly response to different pH environments using a quartz crystal microbalance with dissipation monitoring (QCM-D), swelling studies, ζ potential measurements, and dynamic mechanical analysis (DMA). In an acidic environment, the multilayers presented lower dissipation values and, consequently, higher moduli when compared with the values obtained for the pH used during the assembly (5.5). When the multilayers were exposed to alkaline environments, the opposite behavior occurred. These results were further corroborated by the ability of this multilayered system to exhibit a reversible swelling-deswelling behavior within the pH range from 3 to 9. The changes in the physicochemical properties of the multilayer system were gradual and different from those of individual solubilized polyelectrolytes. This behavior is related to electrostatic interactions between the ionizable groups combined with hydrogen bonding and hydrophobic interactions. Beyond the pH range of 3-9, the multilayers were stabilized by genipin cross-linking. The multilayered films also became more rigid while the pH responsiveness conferred by the ionizable moieties of the polyelectrolytes was preserved. This work demonstrates the versatility and feasibility of LbL methodology to generate inherently pH stimulus-responsive nanostructured films. Surface functionalization using pH responsiveness endows several biomedical applications with abilities such as drug delivery, diagnostics, microfluidics, biosensing, and biomimetic implantable membranes.
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Chitosan, medium molecular weight