Merck
  • IFITM proteins promote SARS-CoV-2 infection and are targets for virus inhibition in vitro.
IFITM proteins promote SARS-CoV-2 infection and are targets for virus inhibition in vitro.
Nature communications (2021-07-30)
Caterina Prelli Bozzo, Rayhane Nchioua, Meta Volcic, Lennart Koepke, Jana Krüger, Desiree Schütz, Sandra Heller, Christina M Stürzel, Dorota Kmiec, Carina Conzelmann, Janis Müller, Fabian Zech, Elisabeth Braun, Rüdiger Groß, Lukas Wettstein, Tatjana Weil, Johanna Weiß, Federica Diofano, Armando A Rodríguez Alfonso, Sebastian Wiese, Daniel Sauter, Jan Münch, Christine Goffinet, Alberto Catanese, Michael Schön, Tobias M Boeckers, Steffen Stenger, Kei Sato, Steffen Just, Alexander Kleger, Konstantin M J Sparrer, Frank Kirchhoff
ABSTRACT
Interferon-induced transmembrane proteins (IFITMs 1, 2 and 3) can restrict viral pathogens, but pro- and anti-viral activities have been reported for coronaviruses. Here, we show that artificial overexpression of IFITMs blocks SARS-CoV-2 infection. However, endogenous IFITM expression supports efficient infection of SARS-CoV-2 in human lung cells. Our results indicate that the SARS-CoV-2 Spike protein interacts with IFITMs and hijacks them for efficient viral infection. IFITM proteins were expressed and further induced by interferons in human lung, gut, heart and brain cells. IFITM-derived peptides and targeting antibodies inhibit SARS-CoV-2 entry and replication in human lung cells, cardiomyocytes and gut organoids. Our results show that IFITM proteins are cofactors for efficient SARS-CoV-2 infection of human cell types representing in vivo targets for viral transmission, dissemination and pathogenesis and are potential targets for therapeutic approaches.
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
apo-Transferrin human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Putrescine dihydrochloride, ≥98% (TLC)