Merck
  • Impact of dietary proteins on energy balance, insulin sensitivity, and glucose homeostasis: From proteins to peptides to amino acids
Impact of dietary proteins on energy balance, insulin sensitivity, and glucose homeostasis: From proteins to peptides to amino acids
The Molecular Nutrition of Amino Acids and Proteins (2016)