Merck
  • The hydrolysis of alpha-D-glucosides by amyloglucosidase from Aspergillus niger.
The hydrolysis of alpha-D-glucosides by amyloglucosidase from Aspergillus niger.
The Journal of biological chemistry (1962-04-01)
J H PAZUR, K KLEPPE
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Roche
Amyloglucosidase, from Aspergillus niger
Sigma-Aldrich
Amyloglucosidase from Rhizopus sp., ≥40,000 units/g solid
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein