Merck
New saccharogenic determination of alpha-amylase in serum and urine.
Clinical chemistry (1979-02-01)
L van Leeuwen
PMID215341
ABSTRACT
I describe a new kinetic enzymatic saccharogenic method for assaying alpha-amylase in human serum and urine. alpha-Amylase liberates maltose from starch. This is successively acted on by alpha-glucosidase, mutarotase, and glucose dehydrogenase. The resulting conversion of NAD+ to NADH, measured at 340 nm, during a 20-min incubation reflects amylase activity. Endogenous glucose is destroyed before measurement of amylase activity is begun.