Merck
  • A Switch from Cell-Associated to Soluble PDGF-B Protects against Atherosclerosis, despite Driving Extramedullary Hematopoiesis.
A Switch from Cell-Associated to Soluble PDGF-B Protects against Atherosclerosis, despite Driving Extramedullary Hematopoiesis.
Cells (2021-08-08)
Renée J H A Tillie, Thomas L Theelen, Kim van Kuijk, Lieve Temmerman, Jenny de Bruijn, Marion Gijbels, Christer Betsholtz, Erik A L Biessen, Judith C Sluimer
ABSTRACT
Platelet-derived growth factor B (PDGF-B) is a mitogenic, migratory and survival factor. Cell-associated PDGF-B recruits stabilizing pericytes towards blood vessels through retention in extracellular matrix. We hypothesized that the genetic ablation of cell-associated PDGF-B by retention motif deletion would reduce the local availability of PDGF-B, resulting in microvascular pericyte loss, microvascular permeability and exacerbated atherosclerosis. Therefore, Ldlr-/-Pdgfbret/ret mice were fed a high cholesterol diet. Although plaque size was increased in the aortic root of Pdgfbret/ret mice, microvessel density and intraplaque hemorrhage were unexpectedly unaffected. Plaque macrophage content was reduced, which is likely attributable to increased apoptosis, as judged by increased TUNEL+ cells in Pdgfbret/ret plaques (2.1-fold) and increased Pdgfbret/ret macrophage apoptosis upon 7-ketocholesterol or oxidized LDL incubation in vitro. Moreover, Pdgfbret/ret plaque collagen content increased independent of mesenchymal cell density. The decreased macrophage matrix metalloproteinase activity could partly explain Pdgfbret/ret collagen content. In addition to the beneficial vascular effects, we observed reduced body weight gain related to smaller fat deposition in Pdgfbret/ret liver and adipose tissue. While dampening plaque inflammation, Pdgfbret/ret paradoxically induced systemic leukocytosis. The increased incorporation of 5-ethynyl-2'-deoxyuridine indicated increased extramedullary hematopoiesis and the increased proliferation of circulating leukocytes. We concluded that Pdgfbret/ret confers vascular and metabolic effects, which appeared to be protective against diet-induced cardiovascular burden. These effects were unrelated to arterial mesenchymal cell content or adventitial microvessel density and leakage. In contrast, the deletion drives splenic hematopoiesis and subsequent leukocytosis in hypercholesterolemia.
Sigma-Aldrich
PDGF-BB from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
REDExtract-N-Amp PCR ReadyMix, Ready-to-use 2X PCR Master Mix with Loading Dye
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests