Merck
  • Antimicrobial and Antioxidant Secondary Metabolites from Trifolium baccarinii Chiov. (Fabaceae) and Their Mechanisms of Antibacterial Action.
Antimicrobial and Antioxidant Secondary Metabolites from Trifolium baccarinii Chiov. (Fabaceae) and Their Mechanisms of Antibacterial Action.
BioMed research international (2021-11-02)
Donald Léonel Feugap Tsamo, Jean-De-Dieu Tamokou, Irene Chinda Kengne, Claudia Darille Jouogo Ngnokam, Mahamat Djamalladine Djamalladine, Laurence Voutquenne-Nazabadioko, David Ngnokam
ABSTRACT
The treatment of infectious diseases with antimicrobial agents continues to present problems in modern-day medicine with many studies showing significant increase in the incidence of bacterial resistance to several antibiotics. The screening of antimicrobial activity of plant extracts and natural products has shown that medicinal plants are made up of a potential source of new anti-infective agents. The aim of this study was to evaluate the antimicrobial and antioxidant activities of extracts and compounds from the whole plant Trifolium baccarinii Chiov. and to determine their modes of antibacterial action. The plant extracts were prepared by maceration in organic solvents. The antimicrobial activities were evaluated using the broth microdilution method. The antioxidant activity was evaluated using the 2,2'-diphenyl-1-picrylhydrazyl radical (DPPH) and 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid) diammonium salt (ABTS) assays. The mechanisms of antibacterial action were determined by lysis, salt tolerance assays, and antioxidant enzyme activities. The cytotoxic effect on the erythrocytes was determined by a spectrophotometric method. Biochanin A, formononetin, luteolin, luteolin-4'-O-β-D-glucopyranoside, 4,7,2'-trihydroxy-4'-methoxyisoflavanol, sissotrin, 1-methyl-β-D-glucopyranoside, ononin, D-mannitol, and 3-O-β-D-glucuronopyranosylsoyasapogenol B were isolated from Trifolium baccarinii. The MeOH, EtOAc, and n-BuOH extracts as well as biochanin A, formononetin, luteolin, luteolin-4'-O-β-D-glucopyranoside, 4,7,2'-trihydroxy-4'-methoxyisoflavanol, and sissotrin from Trifolium baccarinii displayed the highest antimicrobial and antioxidant activities. The MeOH extract and 4,7,2'-trihydroxy-4'-methoxyisoflavanol exhibited antibacterial activity through the bacteriolytic effect and reduction of the antioxidant defenses in the bacterial cells. The present study portrays Trifolium baccarinii as a potential natural source of antibacterial, antifungal, and antioxidant agents.
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.