Merck
  • Shared and Unique Evolutionary Trajectories to Ciprofloxacin Resistance in Gram-Negative Bacterial Pathogens.
Shared and Unique Evolutionary Trajectories to Ciprofloxacin Resistance in Gram-Negative Bacterial Pathogens.
mBio (2021-06-23)
Jaime E Zlamal, Semen A Leyn, Mallika Iyer, Marinela L Elane, Nicholas A Wong, James W Wamsley, Maarten Vercruysse, Fernando Garcia-Alcalde, Andrei L Osterman
ABSTRACT
Resistance to the broad-spectrum antibiotic ciprofloxacin is detected at high rates for a wide range of bacterial pathogens. To investigate the dynamics of ciprofloxacin resistance development, we applied a comparative resistomics workflow for three clinically relevant species of Gram-negative bacteria: Escherichia coli, Acinetobacter baumannii, and Pseudomonas aeruginosa. We combined experimental evolution in a morbidostat with deep sequencing of evolving bacterial populations in time series to reveal both shared and unique aspects of evolutionary trajectories. Representative clone characterization by sequencing and MIC measurements enabled direct assessment of the impact of mutations on the extent of acquired drug resistance. In all three species, we observed a two-stage evolution: (i) early ciprofloxacin resistance reaching 4- to 16-fold the MIC for the wild type, commonly as a result of single mutations in DNA gyrase target genes (gyrA or gyrB), and (ii) additional genetic alterations affecting the transcriptional control of the drug efflux machinery or secondary target genes (DNA topoisomerase parC or parE). IMPORTANCE The challenge of spreading antibiotic resistance calls for systematic efforts to develop more "irresistible" drugs based on a deeper understanding of dynamics and mechanisms of antibiotic resistance acquisition. To address this challenge, we have established a comparative resistomics approach which combines experimental evolution in a continuous-culturing device, the morbidostat, with ultradeep sequencing of evolving microbial populations to identify evolutionary trajectories (mutations and genome rearrangements) leading to antibiotic resistance over a range of target pathogens. Here, we report the comparative resistomics study of three Gram-negative bacteria (Escherichia coli, Acinetobacter baumannii, and Pseudomonas aeruginosa), which revealed shared and species-specific aspects of the evolutionary landscape leading to robust resistance against the clinically important antibiotic ciprofloxacin. Despite some differences between morbidostat-deduced mutation profiles and those observed in clinical isolates of individual species, a cross-species comparative resistomics approach allowed us to recapitulate all types of clinically relevant ciprofloxacin resistance mechanisms. This observation supports the anticipated utility of this approach in guiding rational optimization of treatment regimens for current antibiotics and the development of novel antibiotics with minimized resistance propensities.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 70 purifications