Merck
  • Potent antibody-mediated neutralization limits bacteriophage treatment of a pulmonary Mycobacterium abscessus infection.
Potent antibody-mediated neutralization limits bacteriophage treatment of a pulmonary Mycobacterium abscessus infection.
Nature medicine (2021-07-10)
Rebekah M Dedrick, Krista G Freeman, Jan A Nguyen, Asli Bahadirli-Talbott, Bailey E Smith, Andrew E Wu, Aaron S Ong, Cheng Ting Lin, Lisa C Ruppel, Nicole M Parrish, Graham F Hatfull, Keira A Cohen
ABSTRACT
An 81-year-old immunocompetent patient with bronchiectasis and refractory Mycobacterium abscessus lung disease was treated for 6 months with a three-phage cocktail active against the strain. In this case study of phage to lower infectious burden, intravenous administration was safe and reduced the M. abscessus sputum load tenfold within one month. However, after two months, M. abscessus counts increased as the patient mounted a robust IgM- and IgG-mediated neutralizing antibody response to the phages, which was associated with limited therapeutic efficacy.
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate