Merck
  • Effects of the CLIP fragment ACTH 20-24 on the duration of REM sleep episodes.
Effects of the CLIP fragment ACTH 20-24 on the duration of REM sleep episodes.
Neuropeptides (1997-02-01)
W Wetzel, T Wagner, D Vogel, H U Demuth, D Balschun
ABSTRACT
Substances acting upon rapid eye movement (REM) sleep or paradoxical sleep (PS) can affect the number and/or the duration of PS episodes. In the present study, we investigated the effects of the proopiomelanocortin-derived peptide CLIP (corticotropin-like intermediate lobe peptide, ACTH 18-39) and its N-terminal fragments ACTH 18-24 and ACTH 20-24 on the duration of PS episodes in rats. Intracerebroventricular injection of ACTH 20-24 caused a pronounced prolongation of PS episodes (up to 7 min duration, never seen under baseline conditions), whereas ACTH 18-24 acted in a similar way but without reaching statistical significance. We suggest that short N-terminal CLIP fragment(s) may represent endogenous hypnogenic factor(s) involved in the regulation of paradoxical sleep.
Sigma-Aldrich
ProteoMass ACTH Fragment 18-39 MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 2,464.1989 Da
Sigma-Aldrich
Adrenocorticotropic Hormone Fragment 18-39 human, ≥97% (HPLC)