Merck
  • Effects of L-, D-, and DL-carnitine on morphometric parameters of skeletal muscle and exercise performance of laboratory animals receiving carnitine-deficient diet.
Effects of L-, D-, and DL-carnitine on morphometric parameters of skeletal muscle and exercise performance of laboratory animals receiving carnitine-deficient diet.
Bulletin of experimental biology and medicine (2007-04-07)
A A Spasov, I N Iezhitsa, M S Kravchenko, V B Pisarev, G L Snigur
ABSTRACT
Serum concentration of L-carnitine, the mean thickness of the skeletal muscle fiber, and exercise performance in the forced swimming test decreased in rats receiving a carnitine-deficient diet. Treatment with L-carnitine compensated for carnitine deficiency, while racemate and D-stereoisomer did not increase its level. L-Carnitine, but not racemate and D-stereoisomer, promoted recovery of the skeletal muscle fiber thickness and exercise performance of rats.
Sigma-Aldrich
(±)-Carnitine hydrochloride, ≥98% (TLC)
Pricing and availability is not currently available.