Merck
  • Experimental determination of the gas phase proton affinities of the conjugate base anions of 2-iodoxybenzoic acid (IBX) and 2-iodosobenzoic acid (IBA).
Experimental determination of the gas phase proton affinities of the conjugate base anions of 2-iodoxybenzoic acid (IBX) and 2-iodosobenzoic acid (IBA).
Organic & biomolecular chemistry (2008-07-05)
Tom Waters, Jack Boulton, Timothy Clark, Michael J Gallen, Craig M Williams, Richard A J O'Hair
ABSTRACT
The gas phase anion proton affinities of the periodinane anions IBX- and IBA- were examined using mass spectrometry-based experiments, and estimated as 1300 +/- 25 and 1390 +/- 10 kJ mol(-1), respectively. The experimental results were supported by theoretical calculations, which yielded proton affinities of 1336 and 1392 kJ mol(-1) for IBX- and IBA- respectively, at the B3LYP/aug-cc-PVDZ level of theory.
Sigma-Aldrich
2-Iodosobenzoic acid