Merck
  • D-alaninol adsorption on Cu(100): photoelectron spectroscopy and first-principles calculations.
D-alaninol adsorption on Cu(100): photoelectron spectroscopy and first-principles calculations.
The journal of physical chemistry. B (2008-03-11)
Paola Gori, Giorgio Contini, Tommaso Prosperi, Daniele Catone, Stefano Turchini, Nicola Zema, Amedeo Palma
ABSTRACT
The adsorption of a single molecule of the D-enantiomer of alaninol (2-amino-1-propanol) on the surface of Cu(100) is investigated through density functional theory calculations. Different possible adsorption sites for D-alaninol are tested, and it is found that the most stable configuration presents both amino and hydroxyl group covalently interacting with "on top" copper atoms. The electronic structure is analyzed in detail and compared with experimental photoelectron spectra. Another adsorption structure in which a dehydrogenation process is assumed to occur on the amino group is analyzed and provides a possible explanation of the valence band electronic structure and of the experimentally observed N 1s core-level shift at full coverage, where a self-assembled ordered chiral monolayer is formed on the copper surface.
Sigma-Aldrich
(S)-(+)-2-Amino-1-propanol, 98%
Sigma-Aldrich
DL-Alaninol, 98%