Merck
  • Differential mobility spectrometry of isomeric protonated dipeptides: modifier and field effects on ion mobility and stability.
Differential mobility spectrometry of isomeric protonated dipeptides: modifier and field effects on ion mobility and stability.
Analytical chemistry (2011-04-21)
Voislav Blagojevic, Alexander Chramow, Bradley B Schneider, Thomas R Covey, Diethard K Bohme
ABSTRACT
The ability to resolve isomeric protonated dipeptides was investigated with the new technique of differential ion mobility mass spectrometry that uses "modifier" molecules to enhance differential mobility. Two pairs of protonated peptides [glycine-alanine (GlyAla) and alanine-glycine (AlaGly), glycine-serine (GlySer) and serine-glycine (SerGly)] and eight different modifiers (water, 2-propanol, 1,5-hexadiene, 2-chloropropane, chlorobenzene, dichloromethane, acetonitrile, and cyclohexane) were used in the initial study. Separation of the protonated peptides was found to be dependent on the mass and proton affinity of the modifier and combinations of functionalities present in the modifier and the analyte ion. Six of the eight modifiers (water, 2-propanol, chlorobenzene, cyclohexane, dichloromethane, and acetonitrile) were able to separate the protonated isomeric peptide pairs, and generally, modifiers with electron-rich groups performed the best. In the presence of some modifiers, a reduction of ion current was observed under the highest field conditions (>115 Td). Dopant-catalyzed isomerization, likely by proton-transport catalysis, and field-induced fragmentation may have contributed to these losses. Two high vapor pressure modifiers, 1,5-hexadiene and 2-chloropropane, significantly influenced ion formation leading to the formation of stable cluster populations that could be observed in the mass spectrometer. Although not a major concern, both fragmentation and influence of modifier evaporation warrant further studies in order to fully understand and possibly eliminate them.
Sigma-Aldrich
Ala-Gly
Sigma-Aldrich
Ala-Phe
Sigma-Aldrich
Ala-Leu
Sigma-Aldrich
Ala-Tyr
Sigma-Aldrich
Ala-Pro hydrate