Merck
  • Effects of s-allyl-L-cysteine on cell proliferation and neuroblast differentiation in the mouse dentate gyrus.
Effects of s-allyl-L-cysteine on cell proliferation and neuroblast differentiation in the mouse dentate gyrus.
The Journal of veterinary medical science (2011-04-07)
Sung Min Nam, Dae Young Yoo, Woosuk Kim, Miyoung Yoo, Dong-Woo Kim, Moo-Ho Won, In Koo Hwang, Yeo Sung Yoon
ABSTRACT
In this study, we investigated the effects of S-allyl-L-cysteine (SAC), a major sulfur-containing compound present in garlic, on Ki67- and doublecortin (DCX)-positive cells, which were used as a marker for cell proliferation and neuroblast differentiation, respectively, in the mouse dentate gyrus. SAC (300 mg/kg) was intraperitoneally administered to 8-week-old mice once a day for 3 weeks. The animals were then sacrificed at 11 weeks of age. SAC administration significantly increased Ki67-positive nuclei and DCX-immunoreactive neuroblasts in the subgranular zone of the dentate gyrus. Furthermore, serotonin 1A receptor (5-HT(1A)) levels in the hippocampus were also increased. These results suggest that SAC significantly increased cell proliferation and neuroblast differentiation by increasing 5-HT(1A) levels in the dentate gyrus.
Sigma-Aldrich
(±)-L-Alliin, ≥90% (HPLC)
Sigma-Aldrich
S-Allyl-L-cysteine, ≥98% (HPLC)