Merck
  • Sublethal effects of spirodiclofen, abamectin and pyridaben on life-history traits and life-table parameters of two-spotted spider mite, Tetranychus urticae (Acari: Tetranychidae).
Sublethal effects of spirodiclofen, abamectin and pyridaben on life-history traits and life-table parameters of two-spotted spider mite, Tetranychus urticae (Acari: Tetranychidae).
Experimental & applied acarology (2018-03-10)
Moosa Saber, Zeinab Ahmadi, Gholamreza Mahdavinia
ABSTRACT
Two-spotted spider mite, Tetranychus urticae Koch, is one of the economically most important pests on a wide range of crops in greenhouses and orchards worldwide. Control of T. urticae has been largely based on the use of acaricides. Sublethal effects of spirodiclofen, pyridaben and abamectin were studied on life-table parameters of T. urticae females treated with the acaricides. LC25 values of spirodiclofen, abamectin and pyridaben (3.84, 0.04 and 136.96 µg a.i./ml, respectively) were used for sublethal studies. All acaricides showed significant effects on T. urticae biological parameters including developmental time, survival rate, and fecundity. The females treated with spirodiclofen, abamectin and pyridaben at LC25 exhibited significantly reduced net reproductive rate (R0), finite rate of increase (λ) and intrinsic rate of increase (r). The intrinsic rate of increase in spirodiclofen, abamectin and pyridaben treated groups and control were 0.0138, 0.0273, 0.039 and 0.2481 female offspring per female per day, respectively. The results indicated that sublethal concentrations of tested pesticides strongly affected the life characteristics of spider mite and consequently may influence mite population growth in future generations.
Product Description
Supelco
Spirodiclofen, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.