Merck
  • Insights into the mode of action of a putative zinc transporter CzrB in Thermus thermophilus.
Insights into the mode of action of a putative zinc transporter CzrB in Thermus thermophilus.
Structure (London, England : 1993) (2008-09-13)
Vadim Cherezov, Nicole Höfer, Doletha M E Szebenyi, Olga Kolaj, J Gerard Wall, Richard Gillilan, Vasundara Srinivasan, Christopher P Jaroniec, Martin Caffrey
ABSTRACT
The crystal structures of the cytoplasmic domain of the putative zinc transporter CzrB in the apo and zinc-bound forms reported herein are consistent with the protein functioning in vivo as a homodimer. NMR, X-ray scattering, and size-exclusion chromatography provide support for dimer formation. Full-length variants of CzrB in the apo and zinc-loaded states were generated by homology modeling with the Zn2+/H+ antiporter YiiP. The model suggests a way in which zinc binding to the cytoplasmic fragment creates a docking site to which a metallochaperone can bind for delivery and transport of its zinc cargo. Because the cytoplasmic domain may exist in the cell as an independent, soluble protein, a proposal is advanced that it functions as a metallochaperone and that it regulates the zinc-transporting activity of the full-length protein. The latter requires that zinc binding becomes uncoupled from the creation of a metallochaperone-docking site on CzrB.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Conalbumin from chicken egg white, suitable for cell culture, BioReagent
Pricing and availability is not currently available.
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus
Pricing and availability is not currently available.
Sigma-Aldrich
Conalbumin from chicken egg white, Iron complex
Pricing and availability is not currently available.
Sigma-Aldrich
Conalbumin from chicken egg white, Substantially iron-free
Pricing and availability is not currently available.