Merck
  • Determining the quantitative relationship between glycolysis and GAPDH in cancer cells exhibiting the Warburg effect.
Determining the quantitative relationship between glycolysis and GAPDH in cancer cells exhibiting the Warburg effect.
The Journal of biological chemistry (2021-02-06)
Xiaobing Zhu, Chengmeng Jin, Qiangrong Pan, Xun Hu
ABSTRACT
Previous studies have identified GAPDH as a promising target for treating cancer and modulating immunity because its inhibition reduces glycolysis in cells (cancer cells and immune cells) with the Warburg effect, a modified form of cellular metabolism found in cancer cells. However, the quantitative relationship between GAPDH and the aerobic glycolysis remains unknown. Here, using siRNA-mediated knockdown of GAPDH expression and iodoacetate-dependent inhibition of enzyme activity, we examined the quantitative relationship between GAPDH activity and glycolysis rate. We found that glycolytic rates were unaffected by the reduction of GAPDH activity down to 19% ± 4.8% relative to untreated controls. However, further reduction of GAPDH activity below this level caused proportional reductions in the glycolysis rate. GAPDH knockdown or inhibition also simultaneously increased the concentration of glyceraldehyde 3-phosphate (GA3P, the substrate of GAPDH). This increased GA3P concentration countered the effect of GAPDH knockdown or inhibition and stabilized the glycolysis rate by promoting GAPDH activity. Mechanistically, the intracellular GA3P concentration is controlled by the Gibbs free energy of the reactions upstream of GAPDH. The thermodynamic state of the reactions along the glycolysis pathway was only affected when GAPDH activity was reduced below 19% ± 4.8%. Doing so moved the reactions catalyzed by GAPDH + PGK1 (phosphoglycerate kinase 1, the enzyme immediate downstream of GAPDH) away from the near-equilibrium state, revealing an important biochemical basis to interpret the rate control of glycolysis by GAPDH. Collectively, we resolved the numerical relationship between GAPDH and glycolysis in cancer cells with the Warburg effect and interpreted the underlying mechanism.
Sigma-Aldrich
Hexokinase from Saccharomyces cerevisiae, Type F-300, lyophilized powder, ≥130 units/mg protein (biuret)
Sigma-Aldrich
Enolase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase from rabbit muscle, Type I, ammonium sulfate suspension, 100-300 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type X, lyophilized powder, ≥3,500 units/mg protein
Sigma-Aldrich
Glutathione Reductase from baker′s yeast (S. cerevisiae), ammonium sulfate suspension, 100-300 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Aldolase from rabbit muscle, ammonium sulfate suspension, 10-20 units/mg protein
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
3-Phosphoglyceric Phosphokinase from baker′s yeast (S. cerevisiae), ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)