Merck
  • Chromatin profiling in human neurons reveals aberrant roles for histone acetylation and BET family proteins in schizophrenia.
Chromatin profiling in human neurons reveals aberrant roles for histone acetylation and BET family proteins in schizophrenia.
Nature communications (2022-04-24)
Lorna A Farrelly, Shuangping Zheng, Nadine Schrode, Aaron Topol, Natarajan V Bhanu, Ryan M Bastle, Aarthi Ramakrishnan, Jennifer C Chan, Bulent Cetin, Erin Flaherty, Li Shen, Kelly Gleason, Carol A Tamminga, Benjamin A Garcia, Haitao Li, Kristen J Brennand, Ian Maze
ABSTRACT
Schizophrenia (SZ) is a psychiatric disorder with complex genetic risk dictated by interactions between hundreds of risk variants. Epigenetic factors, such as histone posttranslational modifications (PTMs), have been shown to play critical roles in many neurodevelopmental processes, and when perturbed may also contribute to the precipitation of disease. Here, we apply an unbiased proteomics approach to evaluate combinatorial histone PTMs in human induced pluripotent stem cell (hiPSC)-derived forebrain neurons from individuals with SZ. We observe hyperacetylation of H2A.Z and H4 in neurons derived from SZ cases, results that were confirmed in postmortem human brain. We demonstrate that the bromodomain and extraterminal (BET) protein, BRD4, is a bona fide 'reader' of H2A.Z acetylation, and further provide evidence that BET family protein inhibition ameliorates transcriptional abnormalities in patient-derived neurons. Thus, treatments aimed at alleviating BET protein interactions with hyperacetylated histones may aid in the prevention or treatment of SZ.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid