Merck
Rapid macropinocytic transfer of α-synuclein to lysosomes.
Cell reports (2022-07-21)
Armin Bayati, Emily Banks, Chanshuai Han, Wen Luo, Wolfgang E Reintsch, Cornelia E Zorca, Irina Shlaifer, Esther Del Cid Pellitero, Benoit Vanderperre, Heidi M McBride, Edward A Fon, Thomas M Durcan, Peter S McPherson
ABSTRACT
The nervous system spread of alpha-synuclein fibrils is thought to cause Parkinson's disease (PD) and other synucleinopathies; however, the mechanisms underlying internalization and cellular spread are enigmatic. Here, we use confocal and superresolution microscopy, subcellular fractionation, and electron microscopy (EM) of immunogold-labeled α-synuclein preformed fibrils (PFFs) to demonstrate that this form of the protein undergoes rapid internalization and is targeted directly to lysosomes in as little as 2 min. Uptake of PFFs is disrupted by macropinocytic inhibitors and circumvents classical endosomal pathways. Immunogold-labeled PFFs are seen at the highly curved inward edge of membrane ruffles, in newly formed macropinosomes, in multivesicular bodies and in lysosomes. While most fibrils remain in lysosomes, a portion is transferred to neighboring naive cells along with markers of exosomes. These data indicate that PFFs use a unique internalization mechanism as a component of cell-to-cell propagation.
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, Magnetic bead Conjugate, purified antibody, clone 23A8, from mouse
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture