Merck
  • Deficiency of peroxisomal NUDT7 stimulates de novo lipogenesis in hepatocytes.
Deficiency of peroxisomal NUDT7 stimulates de novo lipogenesis in hepatocytes.
iScience (2022-10-04)
Jinsoo Song, In-Jeoung Baek, Sujeong Park, Jinjoo Oh, Deokha Kim, Kyung Song, Mi Kyung Kim, Hye Won Lee, Byoung Kuk Jang, Eun-Jung Jin
ABSTRACT
Here, we found that heterozygous null of peroxisomal Nudt7 (Nudt7 +/- ) induced the typical NAFLD features, i.e. increased levels of hepatic triglyceride (TG) and fatty acid (FA), infiltration of inflammatory cells, impaired glucose tolerance and insulin sensitivity, and stimulation of lipolysis from adipose tissue. Particularly, in Nudt7 +/- hepatocytes, de novo lipogenesis (DNL) was significantly increased. Ingenuity pathway analysis (IPA) and KEGG pathway analysis of RNA sequencing data suggested the activation of PPAR signaling in the liver of Nudt7 +/- mice. Moreover, accumulation of palmitic acid in Nudt7 +/- hepatocyte increased the level of H3K4me3 on the promoters of PPARγ resulting in the activation of PPARγ and induced the DNL in the hepatocytes of Nudt7 +/- mice. Moreover, we found that liraglutide significantly reduced typical NAFLD features induced by NUDT7 deficiency. Our data suggest that dysregulation of peroxisomal NUDT7 is responsible for upregulation of hepatic DNL by accumulation of palmitic acid and PPARγ activation.
Sigma-Aldrich
Filipin complex ready made solution, from Streptomyces filipinensis, 5mg/ml in DMSO based solution
Pricing and availability is not currently available.