Merck
  • Engraftable Induced Pluripotent Stem Cell-Derived Neural Precursors for Brain Repair.
Engraftable Induced Pluripotent Stem Cell-Derived Neural Precursors for Brain Repair.
Methods in molecular biology (Clifton, N.J.) (2020-06-01)
Ourania Zygogianni, Georgia Kouroupi, Era Taoufik, Rebecca Matsas
ABSTRACT
Stem cell transplantation has attracted great interest for treatment of neurodegenerative diseases to provide neuroprotection, repair the lesioned neuronal network and restore functionality. Parkinson's disease (PD), in particular, has been a preferred target because motor disability that constitutes a core pathology of the disease is associated with local loss of dopaminergic neurons in a specific brain area, the substantia nigra pars compacta. These cells project to the striatum where they deliver the neurotransmitter dopamine that is involved in control of many aspects of motor behavior. Therefore, cell transplantation approaches in PD aim to replenish dopamine deficiency in the striatum. A major challenge in developing cell therapy approaches is the ability to generate large numbers of transplantable cells in a reliable and reproducible manner. In recent years the technological breakthrough of induced pluripotent stem cells (iPSCs) has demonstrated that this is possible at a preclinical level, accelerating clinical translation. A second important issue is to efficiently differentiate iPSCs into dopaminergic neuronal progenitors with restricted proliferation potential in order to avoid cellular overgrowth in vivo and minimize the risk of tumorigenesis. Here we describe an effective protocol that includes human iPSC differentiation to the dopaminergic lineage and enrichment in neuronal precursor cells expressing the polysialylated form of the neural cell adhesion molecule PSA-NCAM, through magnetically activated cell sorting. The resulting cells are transplanted and shown to survive, differentiate, and integrate within a striatal lesion model generated by unilateral 6-hydroxydopamine administration in mice of the NOD/SCID strain that supports xenografts.
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit