Merck
  • Immunohistochemical localization of novel CART peptides in rat hypothalamus, pituitary and adrenal gland.
Immunohistochemical localization of novel CART peptides in rat hypothalamus, pituitary and adrenal gland.
Journal of neuroendocrinology (1998-01-07)
E O Koylu, P R Couceyro, P D Lambert, N C Ling, E B DeSouza, M J Kuhar
ABSTRACT
CART peptide specific polyclonal antisera were raised in rabbits. The antisera were raised to CART peptide fragments that span most of the predicted CART protein. The specificity of each antisera was demonstrated by blockade of immunostaining by the immunizing peptide but not by the other CART peptide fragments. In the hypothalamus and pituitary of colchicine and noncolchicine treated rats, immunostaining was observed in cell bodies, fibers and varicosities. Clusters of cells were also stained in the adrenal medulla. It is noteworthy that cellular immunostaining was only found in areas previously shown to express CART mRNA. These findings indicate the presence of CART peptide(s) in the hypothalamus, pituitary, and adrenal gland. Furthermore, we also present evidence for the possible processing of the CART pro-peptide into smaller peptide fragments. These neuroanatomical findings suggest a role of CART peptides in hypothalamic, pituitary and adrenal function.