Merck
  • Analysis of diagnostic performance and factors causing nonspecific reactions in SARS-CoV-2 rapid antigen detection tests.
Analysis of diagnostic performance and factors causing nonspecific reactions in SARS-CoV-2 rapid antigen detection tests.
Journal of infection and chemotherapy : official journal of the Japan Society of Chemotherapy (2022-10-27)
Natsuki Narumi, Takashi Kondo, Yuki Sato, Yuki Katayama, Shinya Nirasawa, Masachika Saeki, Yuki Yakuwa, Yoshihiro Fujiya, Koji Kuronuma, Satoshi Takahashi
ABSTRACT
Early diagnosis and appropriate infection control are important to prevent the transmission of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). In this study, we aimed to assess the diagnostic performance of SARS-CoV-2 rapid antigen detection (RAD) tests and the factors that cause nonspecific reactions. Nasopharyngeal swab specimens (n = 100), sputum specimens (n = 10), and lithium-heparin plasma samples (n = 100) were collected. We evaluated Espline®SARS-CoV-2 (Espline) and SARS-CoV-2 Rapid Antigen Test that also known as STANDARD Q® (STANDARD Q), with reverse transcription-polymerase chain reaction (RT-PCR) and Lumipulse® Presto SARS-CoV-2 Ag as reference tests. In addition, we investigated the effects of inadequate pretreatment methods and five potential causes of nonspecific reactions. The sensitivities of Espline and STANDARD Q were 60% and 57%, respectively, and their specificity was 100%. It was confirmed that the judgment line for the positive insufficiently mixed specimens was faint. A false-positive result was observed with STANDARD Q when sputum was used as a specimen to investigate judgment the effect of viscosity. Espline and STANDARD Q show good sensitivity for specimens with Ct values less than 25, but specimens collected within 9 days of symptom onset may still give false negatives. The test should be performed carefully, and the results should be judged comprehensively, taking into account clinical symptoms and patient background.
Sigma-Aldrich
Mucin from porcine stomach, Type II