Merck
  • Self-assembly of pericentriolar material in interphase cells lacking centrioles.
Self-assembly of pericentriolar material in interphase cells lacking centrioles.
eLife (2022-07-06)
Fangrui Chen, Jingchao Wu, Malina K Iwanski, Daphne Jurriens, Arianna Sandron, Milena Pasolli, Gianmarco Puma, Jannes Z Kromhout, Chao Yang, Wilco Nijenhuis, Lukas C Kapitein, Florian Berger, Anna Akhmanova
ABSTRACT
The major microtubule-organizing center (MTOC) in animal cells, the centrosome, comprises a pair of centrioles surrounded by pericentriolar material (PCM), which nucleates and anchors microtubules. Centrosome assembly depends on PCM binding to centrioles, PCM self-association and dynein-mediated PCM transport, but the self-assembly properties of PCM components in interphase cells are poorly understood. Here, we used experiments and modeling to study centriole-independent features of interphase PCM assembly. We showed that when centrioles are lost due to PLK4 depletion or inhibition, dynein-based transport and self-clustering of PCM proteins are sufficient to form a single compact MTOC, which generates a dense radial microtubule array. Interphase self-assembly of PCM components depends on γ-tubulin, pericentrin, CDK5RAP2 and ninein, but not NEDD1, CEP152, or CEP192. Formation of a compact acentriolar MTOC is inhibited by AKAP450-dependent PCM recruitment to the Golgi or by randomly organized CAMSAP2-stabilized microtubules, which keep PCM mobile and prevent its coalescence. Linking of CAMSAP2 to a minus-end-directed motor leads to the formation of an MTOC, but MTOC compaction requires cooperation with pericentrin-containing self-clustering PCM. Our data reveal that interphase PCM contains a set of components that can self-assemble into a compact structure and organize microtubules, but PCM self-organization is sensitive to motor- and microtubule-based rearrangement.
Sigma-Aldrich
Anti-CEP135 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution