Merck
  • Cancer-associated fibroblast-derived exosomal microRNA-20a suppresses the PTEN/PI3K-AKT pathway to promote the progression and chemoresistance of non-small cell lung cancer.
Cancer-associated fibroblast-derived exosomal microRNA-20a suppresses the PTEN/PI3K-AKT pathway to promote the progression and chemoresistance of non-small cell lung cancer.
Clinical and translational medicine (2022-07-21)
Lin Shi, Weiliang Zhu, Yuanyuan Huang, Lin Zhuo, Siyun Wang, Shaobing Chen, Bei Zhang, Bin Ke
ABSTRACT
Cancer-associated fibroblasts (CAFs) contributes to overall tumor progression. In the current survey, we explored the ability of microRNA-20a (miR-20a) within these CAF-derived exosomes to influence non-small-cell lung cancer (NSCLC) progression. Normal tissue-associated fibroblasts (NAFs) and CAFs were collected from samples of NSCLC patient tumors and paracancerous lung tissues. Exosomes derived from these cells were then characterized via Western blotting, nanoparticle tracking analyses, and transmission electron microscopy. The expression of miR-20a was assessed via qPCR and fluorescence in situ hybridization (FISH). CCK-8, EdU uptake, and colony formation assessments were used for evaluating tumor proliferation, while Hoechst staining was performed to monitor the in vitro apoptotic death of tumor cells. A model of xenograft tumor established in nude mice was also used to evaluate in vivo tumor responses. CAF-derived exosomes exhibited miR-20a upregulation and promoted NSCLC cell proliferation and resistance to cisplatin (DDP). Mechanistically, CAF-derived exosomes were discovered to transmit miR-20a to tumor cells wherein it was able to target PTEN to enhance DDP resistance and proliferation. Associated PTEN downregulation following exosome-derived miR-20a treatment enhanced PI3K/AKT pathway activation. The achieved outcomes explain that CAFs can release miR-20a-containing exosomes capable of promoting NSCLC progression and chemoresistance, highlighting this pathway as a possible therapeutic target in NSCLC.
Sigma-Aldrich
Collagenase type I, The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.
Hyaluronidase, European Pharmacopoeia (EP) Reference Standard