Merck
  • Efficient expression, purification, and visualization by cryo-EM of unliganded near full-length HER3.
Efficient expression, purification, and visualization by cryo-EM of unliganded near full-length HER3.
Methods in enzymology (2022-05-08)
Devan Diwanji, Raphael Trenker, Natalia Jura, Kliment A Verba
ABSTRACT
Biochemical analyses of membrane receptor kinases have been limited by challenges in obtaining sufficient homogeneous receptor samples for downstream structural and biophysical characterization. Here, we report a suite of methods for the efficient expression, purification, and visualization by cryo-electron microscopy (cryo-EM) of near full-length Human Epidermal Growth Factor Receptor 3 (HER3), a receptor tyrosine pseudokinase, in the unliganded state. Through transient mammalian cell expression, a two-step purification with detergent exchange into lauryl maltose neopentyl glycol (LMNG), and freezing devoid of background detergent micelle, we obtained ~6Å reconstructions of the ~60kDa fully-glycosylated unliganded extracellular domain of HER3 from just 30mL of suspension culture. The reconstructions reveal previously unappreciated extracellular domain dynamics and glycosylation sites.
Product Description
Millipore
EDTA, Disodium Salt, Dihydrate, Molecular Biology Grade
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)