Merck
  • Regulation and functional impact of lipopolysaccharide induced Nod2 gene expression in the murine epididymal epithelial cell line PC1.
Regulation and functional impact of lipopolysaccharide induced Nod2 gene expression in the murine epididymal epithelial cell line PC1.
Immunology (2008-02-21)
Marcus Mühlbauer, Adam W Cheely, Suresh Yenugu, Christian Jobin
ABSTRACT
Epididymitis represents a serious threat to male fertility and usually develops following secondary bacterial infection of the epididymis such as urinary tract infections or sexually transmitted diseases. Surprisingly, very little is known about the innate host response triggered by bacterial infection in the male reproductive tract. In this study we investigated the regulation and function of Nod2 in epididymal epithelial cells following lipopolysaccharide (LPS) stimulation. The immortalized epididymal epithelial cell line PC1 (proximal caput 1) constitutively expressed Toll-like receptor 4, MD-2, CD-14 but not Nod2 messenger RNA. Lipopolysaccharide (LPS; 0.5 microg/ml) rapidly induced I kappaB phosphorylation and degradation, RelA nuclear translocation and phosphorylation, which correlated with enhanced transcriptional activity (four-fold) in PC1 cells. The LPS and lipid A rapidly (1 hr) induced Nod2 messenger RNA accumulation in a dose-dependent manner. RelA and RNApolII recruitment to the Nod2 gene promoter was enhanced in LPS-stimulated cells. Molecular blockade of nuclear factor-kappaB signalling with adenovirus 5 (Ad5) I kappaB AA or adenovirus 5 double-negative (Ad5dn) IKK beta prevented LPS-induced Nod2 gene expression. Functionally, Nod2 upregulation enhanced muramyl dipeptide (MDP) -induced tumour necrosis factor messenger RNA accumulation in PC1 cells. We conclude that epididymal epithelial cells mount an innate response following LPS exposure which leads to upregulation of Nod2 and enhanced responsiveness to the microbial product MDP. The rapid Nod2 upregulation in epididymal epithelial cells is probably part of a complex innate host response aimed at protecting the male reproductive tract from the deleterious impact of bacteria.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, FITC conjugate
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, Detoxified
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by trichloroacetic acid extraction