Merck
  • Integrated Direct-to-Biology Platform for the Nanoscale Synthesis and Biological Evaluation of PROTACs.
Integrated Direct-to-Biology Platform for the Nanoscale Synthesis and Biological Evaluation of PROTACs.
Journal of medicinal chemistry (2023-11-07)
Rebecca Stevens, Enrique Bendito-Moll, David J Battersby, Afjal H Miah, Natalie Wellaway, Robert P Law, Peter Stacey, Diana Klimaszewska, Justyna M Macina, Glenn A Burley, John D Harling
ABSTRACT
Proteolysis targeting chimeras (PROTACs) are heterobifunctional molecules that co-opt the cell's natural proteasomal degradation mechanisms to degrade undesired proteins. A challenge associated with PROTACs is the time and resource-intensive optimization; thus, the development of high-throughput platforms for their synthesis and biological evaluation is required. In this study, we establish an ultra-high-throughput experimentation (ultraHTE) platform for PROTAC synthesis, followed by direct addition of the crude reaction mixtures to cellular degradation assays without any purification. This 'direct-to-biology' (D2B) approach was validated and then exemplified in a medicinal chemistry campaign to identify novel BRD4 PROTACs. Using the D2B platform, the synthesis of 650 PROTACs was carried out in a 1536-well plate, and subsequent biological evaluation was performed by a single scientist in less than 1 month. Due to its ability to hugely accelerate the optimization of new degraders, we anticipate our platform will transform the synthesis and testing of PROTACs.
Sigma-Aldrich
QuicTPD Acid Screening Set
Sigma-Aldrich
QuicTPD Amine Optimization Set
Sigma-Aldrich
QuicTPD Acid Optimization Set
Sigma-Aldrich
QuicTPD Amine Screening Set