Merck
  • Effect of sex steroids and insulin on dehydroepiandrosterone sulfate production by hepatoma G2 cells.
Effect of sex steroids and insulin on dehydroepiandrosterone sulfate production by hepatoma G2 cells.
Fertility and sterility (2008-06-17)
Marita Pall, Margaret Nguyen, Denis Magoffin, Ricardo Azziz
ABSTRACT
To test the hypothesis that DHEAS production from DHEA occurs in hepatic cells and that this production is augmented by the presence of sex steroids or insulin. In vitro prospective experiment. Academic medical center. Hepatoma G2 cells cultured in media supplemented with [1] DHEA (10(-5) mol/L) only, [2] DHEA (10(-5) mol/L) + T (10(-6) mol/L), [3] DHEA (10(-5) mol/L) + E(2) (10(-6) mol/L), [4] DHEA (10(-5) mol/L) + dihydrotestosterone (10(-6) mol/L), [5] DHEA (10(-5) mol/L) + insulin (10 ng/mL), or [6] DHEA (10(-5) mol/L) + insulin (100 ng/mL). Levels of DHEAS in the media were measured at 0, 2, 4, 6, 8, 12, 24, 48, and 72 hours after adding treatments at time-point 0. Dehydroepiandrosterone sulfate was first detected in the hepatoma G2 cell culture media at 12 hours of incubation. The cumulative production rate of DHEAS increased linearly until 72 hours of incubation. When compared with the effect of treatment with DHEA only, treatment with DHEA plus T, dihydrotestosterone, or E(2) delayed the cumulative DHEAS production; alternatively, the addition of insulin did not alter DHEAS production. These data suggest that although hepatic cells have the ability of converting DHEA to DHEAS, neither sex steroids nor insulin results in the increased hepatic production of DHEAS.
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra