Merck
  • Porcine embryos produced after intracytoplasmic sperm injection using xenogeneic pig sperm from neonatal testis tissue grafted in mice.
Porcine embryos produced after intracytoplasmic sperm injection using xenogeneic pig sperm from neonatal testis tissue grafted in mice.
Reproduction, fertility, and development (2008-10-10)
Ali Honaramooz, Xiang-Shun Cui, Nam-Hyung Kim, Ina Dobrinski
ABSTRACT
Embryo development after homologous intracytoplasmic sperm injection (ICSI) with sperm from testis tissue xenografts from pigs or any other farm animal species has not been evaluated critically. Here, we report development of porcine embryos in vitro following ICSI with sperm retrieved from xenografted neonatal pig testis. Small pieces of testis tissue from newborn piglets were grafted under the back skin of castrated immunodeficient mice (n = 4) and the xenografts were collected 8 months after grafting. Spermatozoa were recovered by mincing of the grafted tissue. For comparison, testicular, epididymal and ejaculated spermatozoa were also collected from mature boars. Oocytes injected with xenogeneic spermatozoa were either fixed to determine fertilisation processes (n = 89 in five replicates) or allowed to develop in vitro (n = 143 in four replicates). Xenogeneic porcine spermatozoa were fertilisation competent (24% v. 58%, 68%, 62% or 0% for xenogeneic v. control testicular, epididymal and ejaculated spermatozoa or no spermatozoa, respectively) and embryos developed to the blastocyst stage (8% v. 22%, 27%, 25% or 0%, respectively). These results demonstrate that porcine spermatozoa derived from immature testis tissue xenografted into mice are fertilisation competent, albeit at a lower rate than testicular, epididymal or ejaculated spermatozoa from control boars, and support embryo development after ICSI.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)