Merck
  • Very small embryonic-like stem cells are present in adult murine organs: ImageStream-based morphological analysis and distribution studies.
Very small embryonic-like stem cells are present in adult murine organs: ImageStream-based morphological analysis and distribution studies.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2008-10-28)
Ewa K Zuba-Surma, Magdalena Kucia, Wan Wu, Izabela Klich, James W Lillard, Janina Ratajczak, Mariusz Z Ratajczak
ABSTRACT
Recently, we purified a population of CXCR4+/Oct-4+/SSEA-1+/Sca-1+/Lin(-)/CD45(-) very small embryonic-like stem cells (VSELs) from adult murine bone marrow (BM). After using flow cytometry, ImageStream analysis, confocal microscopy, and real time RT-PCR, we report that similar cells could be also identified and isolated from several organs in adult mice. The highest total numbers of Oct-4+ VSELs were found in the brain, kidneys, muscles, pancreas, and BM. These observations support our hypothesis that a population of very primitive cells expressing germ line/epiblast markers (Oct-4, SSEA-1) is deposited early during embryogenesis in various organs and survives into adulthood. Further studies are needed to determine whether these cells, after being isolated from various adult human organs similarly to their murine BM-derived counterparts, are endowed with pluripotent stem cell properties.
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free